MAN182109279 Manchester Organics

Ethyl 3-indazolecarboxylate

  • MDL number MFCD01138134
  • Molecular Weight : 190.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T18915

Safety Information

MSDS https://www.manchesterorganics.com/T18915
Availability details for
Quantity
  • Availability
  • Add to Cart