MAN182109282 Manchester Organics

3-Chloro-2-fluorobenzotrifluoride

  • MDL number MFCD11100532
  • Cas Number : 1099597-93-9
  • Molecular Weight : 198.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-chloro-2-fluoro-3-(trifluoromethyl)benzene
Inchi Code 1S/C7H3ClF4/c8-5-3-1-2-4(6(5)9)7(10,11)12/h1-3H
Inchi Key XBBZLOUANPLRIR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/H15049
Availability details for
Quantity
  • Availability
  • Add to Cart