MAN182109286 Manchester Organics

5-(Trifluoromethyl)isoquinoline

  • MDL number MFCD11100536
  • Molecular Weight : 197.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N15054
Availability details for
Quantity
  • Availability
  • Add to Cart