MAN182109300 Manchester Organics

4-(2-Bromoethyl)benzoic acid

  • MDL number MFCD00016541
  • Cas Number : 52062-92-7
  • Molecular Weight : 229.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2-bromoethyl)benzoic acid
Inchi Code 1S/C9H9BrO2/c10-6-5-7-1-3-8(4-2-7)9(11)12/h1-4H,5-6H2,(H,11,12)
Inchi Key BKMRWJWLBHHGCF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y18684
Availability details for
Quantity
  • Availability
  • Add to Cart