MAN182109304 Manchester Organics

3-Aminopyridine-2-carboxylic acid

  • MDL number MFCD00090153
  • Molecular Weight : 138.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L17938

Safety Information

MSDS https://www.manchesterorganics.com/L17938
Availability details for
Quantity
  • Availability
  • Add to Cart