MAN182109314 Manchester Organics

5-Amino-1-(4-bromo-phenyl)-1H-pyrazole-4-carboxylic acid ethyl ester

  • MDL number MFCD03934811
  • Molecular Weight : 310.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L18790

Safety Information

MSDS https://www.manchesterorganics.com/L18790
Availability details for
Quantity
  • Availability
  • Add to Cart