MAN182109317 Manchester Organics

5-Amino-1-(4-chloro-phenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD00973906
  • Molecular Weight : 237.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P18864

Safety Information

MSDS https://www.manchesterorganics.com/P18864
Availability details for
Quantity
  • Availability
  • Add to Cart