MAN182109333 Manchester Organics

2-Methyl-5-(trifluoromethyl)pyridine

  • MDL number MFCD11100538
  • Molecular Weight : 161.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P15030
Availability details for
Quantity
  • Availability
  • Add to Cart