MAN182109352 Manchester Organics

2-Chloro-3-fluorobenzotrifluoride

  • MDL number MFCD11100537
  • Cas Number : 109957-97-3
  • Molecular Weight : 198.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/N15077
Availability details for
Quantity
  • Availability
  • Add to Cart