MAN182109372 Manchester Organics

5-Bromo-2-chloro-4-(trifluoromethyl)pyridine

  • MDL number MFCD11100520
  • Molecular Weight : 260.439

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A15088
Availability details for
Quantity
  • Availability
  • Add to Cart