MAN182109375 Manchester Organics

2-Chloro-4,5-bis-(trifluoromethyl)pyridine

  • MDL number MFCD00082556
  • Cas Number : 109919-25-7
  • Molecular Weight : 249.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/D15091
Availability details for
Quantity
  • Availability
  • Add to Cart