MAN182109376 Manchester Organics

Methyl 2-methylnicotinate

  • MDL number MFCD00051961
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G17911
Availability details for
Quantity
  • Availability
  • Add to Cart