MAN182109399 Manchester Organics

Methyl 1-BOC-3-pyrrolidine carboxylate

  • MDL number MFCD04038683
  • Cas Number : 122684-33-7
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-tert-butyl 3-methyl 1,3-pyrrolidinedicarboxylate
Inchi Code 1S/C11H19NO4/c1-11(2,3)16-10(14)12-6-5-8(7-12)9(13)15-4/h8H,5-7H2,1-4H3
Inchi Key LIWFYAVKYUQMRE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/D18737
Availability details for
Quantity
  • Availability
  • Add to Cart