MAN182109403 Manchester Organics

4-Methyl-2-(trifluoromethyl)pyrimidine-5-carboxylic acid

  • MDL number MFCD11100557
  • Cas Number : 306960-74-7
  • Molecular Weight : 206.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/M10935
Availability details for
Quantity
  • Availability
  • Add to Cart