MAN182109404 Manchester Organics

5-Chloro-3-methylbenzo[b]thiophene- 2-sulfonamide

  • MDL number MFCD00210142
  • Cas Number : 175203-94-8
  • Molecular Weight : 261.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-3-methyl-1-benzothiophene-2-sulfonamide
Inchi Code 1S/C9H8ClNO2S2/c1-5-7-4-6(10)2-3-8(7)14-9(5)15(11,12)13/h2-4H,1H3,(H2,11,12,13)
Inchi Key CGMQMZKLIJYAKX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/H15095
Availability details for
Quantity
  • Availability
  • Add to Cart