MAN182109406 Manchester Organics

5-Cyanopyrimidine

  • MDL number MFCD00235052
  • Molecular Weight : 105.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J18813

Safety Information

MSDS https://www.manchesterorganics.com/J18813
Availability details for
Quantity
  • Availability
  • Add to Cart