MAN182109418 Manchester Organics

6-Fluoroisatin

  • MDL number MFCD03618556
  • Cas Number : 324-03-8
  • Molecular Weight : 165.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/X18754
Availability details for
Quantity
  • Availability
  • Add to Cart