MAN182109419 Manchester Organics

1,2,3,4-Tetrahydro-1-naphthoic acid

  • MDL number MFCD02179127
  • Molecular Weight : 176.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z18756
Availability details for
Quantity
  • Availability
  • Add to Cart