MAN182109452 Manchester Organics

2-Methyloxazole-5-carboxylic acid methyl ester

  • MDL number MFCD06738687
  • Cas Number : 651059-70-0
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/W18776
Availability details for
Quantity
  • Availability
  • Add to Cart