MAN182109460 Manchester Organics

Methyl 2-aminooxazole-5-carboxylate

  • MDL number MFCD09265526
  • Molecular Weight : 142.113

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E18784
Availability details for
Quantity
  • Availability
  • Add to Cart