MAN182109467 Manchester Organics

3,5-Dichloro-2-(trifluoromethyl)pyridine

  • MDL number MFCD11226518
  • Molecular Weight : 215.98798

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T15104

Safety Information

MSDS https://www.manchesterorganics.com/T15104
Availability details for
Quantity
  • Availability
  • Add to Cart