MAN182109482 Manchester Organics

4-Fluoro-2-methoxybenzonitrile

  • MDL number MFCD04116335
  • Cas Number : 191014-55-8
  • Molecular Weight : 151.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/R18794
Availability details for
Quantity
  • Availability
  • Add to Cart