MAN182109487 Manchester Organics

3-Aminophthalimide

  • MDL number MFCD00041853
  • Cas Number : 2518-24-3
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/V18798
Availability details for
Quantity
  • Availability
  • Add to Cart