MAN182109517 Manchester Organics

2-Bromo-N-(4-chlorobenzyl)-acetamide

  • MDL number MFCD11226535
  • Cas Number : 934185-00-9
  • Molecular Weight : 262.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-N-(4-chlorobenzyl)acetamide
Inchi Code 1S/C9H9BrClNO/c10-5-9(13)12-6-7-1-3-8(11)4-2-7/h1-4H,5-6H2,(H,12,13)
Inchi Key HFWRJHHLKNQBHJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/K10933
Availability details for
Quantity
  • Availability
  • Add to Cart