MAN182109520 Manchester Organics

3-Amino-2,6-difluorobenzonitrile

  • MDL number MFCD00120310
  • Molecular Weight : 154.11686

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X18825

Safety Information

MSDS https://www.manchesterorganics.com/X18825
Availability details for
Quantity
  • Availability
  • Add to Cart