MAN182109544 Manchester Organics

2-Methyl-5-(trifluoromethyl)benzaldehyde

  • MDL number MFCD06660200
  • Cas Number : 886498-85-7
  • Molecular Weight : 188.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/J18836
Availability details for
Quantity
  • Availability
  • Add to Cart