MAN182109563 Manchester Organics

5-Chloro-2-methylbenzotrifluoride

  • MDL number MFCD09037961
  • Cas Number : 1839-92-5
  • Molecular Weight : 194.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-1-methyl-2-(trifluoromethyl)benzene
Inchi Code 1S/C8H6ClF3/c1-5-2-3-6(9)4-7(5)8(10,11)12/h2-4H,1H3
Inchi Key WGBKESSICUWUHH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/X15133
Availability details for
Quantity
  • Availability
  • Add to Cart