MAN182109576 Manchester Organics

1-(2,2,2-Trifluoroethyl)-3-(trifluoromethyl)benzene

  • MDL number MFCD11226548
  • Molecular Weight : 228.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/D15139
Availability details for
Quantity
  • Availability
  • Add to Cart