MAN238928520 Manchester Organics

Ethyl 1H-indazole-4-carboxylate

  • MDL number MFCD07371611
  • Cas Number : 885279-45-8
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 1H-indazole-4-carboxylate
Inchi Code 1S/C10H10N2O2/c1-2-14-10(13)7-4-3-5-9-8(7)6-11-12-9/h3-6H,2H2,1H3,(H,11,12)
Inchi Key ZYDRWPMGDIWCRN-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/J18859
Availability details for
Quantity
  • Availability
  • Add to Cart