MAN238928539 Manchester Organics

3-(1,1-Difluoropropyl)pyridine

  • MDL number MFCD11226592
  • Cas Number : 1186194-56-8
  • Molecular Weight : 157.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/N15171
Availability details for
Quantity
  • Availability
  • Add to Cart