MAN238928548 Manchester Organics

2-Phenyl-4-(trifluoromethyl)quinoline

  • MDL number MFCD11226599
  • Molecular Weight : 273.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X15701

Safety Information

MSDS https://www.manchesterorganics.com/X15701
Availability details for
Quantity
  • Availability
  • Add to Cart