MAN238928620 Manchester Organics

4-Methoxy-3,5-difluoropyridine

  • MDL number MFCD12024852
  • Molecular Weight : 145.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B19491

Safety Information

MSDS https://www.manchesterorganics.com/B19491
Availability details for
Quantity
  • Availability
  • Add to Cart