MAN238928633 Manchester Organics

2,3-Diamino-6-methoxypyridine dihydrochloride

  • MDL number MFCD00137799
  • Molecular Weight : 212.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E16157

Safety Information

MSDS https://www.manchesterorganics.com/E16157
Availability details for
Quantity
  • Availability
  • Add to Cart