MAN238928643 Manchester Organics

2-Benzothiazole-2-oxyacetic acid

  • MDL number MFCD07368969
  • Molecular Weight : 209.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F17319

Safety Information

MSDS https://www.manchesterorganics.com/F17319
Availability details for
Quantity
  • Availability
  • Add to Cart