MAN238928652 Manchester Organics

4-Fluoro-1,2-bis-(trifluoromethyl)benzene

  • MDL number MFCD11100160
  • Cas Number : 1092349-75-1
  • Molecular Weight : 232.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/M14011
Availability details for
Quantity
  • Availability
  • Add to Cart