MAN238928659 Manchester Organics

1-Bromo-4-chloro-2-(2,2,2-trifluoroethyl)benzene

  • MDL number MFCD11226573
  • Molecular Weight : 273.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P14013
Availability details for
Quantity
  • Availability
  • Add to Cart