MAN238928667 Manchester Organics

2,3-Dimethylbenzotrifluoride

  • MDL number MFCD11226579
  • Molecular Weight : 174.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/V14018
Availability details for
Quantity
  • Availability
  • Add to Cart