MAN238928671 Manchester Organics

1-Fluoro-3-(2,2,2-trifluoroethyl)benzene

  • MDL number MFCD11100171
  • Cas Number : 81577-08-4
  • Molecular Weight : 178.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-fluoro-3-(2,2,2-trifluoroethyl)benzene
Inchi Code 1S/C8H6F4/c9-7-3-1-2-6(4-7)5-8(10,11)12/h1-4H,5H2
Inchi Key XEMUQSFXYQZZSO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z14022
Availability details for
Quantity
  • Availability
  • Add to Cart