MAN238928672 Manchester Organics

1,3-Dichloro-2-(2,2,2-trifluoroethyl)benzene

  • MDL number MFCD11226581
  • Cas Number : 1099598-25-0
  • Molecular Weight : 229.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-dichloro-2-(2,2,2-trifluoroethyl)benzene
Inchi Code 1S/C8H5Cl2F3/c9-6-2-1-3-7(10)5(6)4-8(11,12)13/h1-3H,4H2
Inchi Key IWVCYQDSZUAMEP-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/A14023
Availability details for
Quantity
  • Availability
  • Add to Cart