MAN238928681 Manchester Organics

1-(2-Amino-1,3-thiazol-5-yl)ethanone

  • MDL number MFCD00775044
  • Molecular Weight : 142.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J18339

Safety Information

MSDS https://www.manchesterorganics.com/J18339
Availability details for
Quantity
  • Availability
  • Add to Cart