MAN238928692 Manchester Organics

1,7-Dimethylindazole-5-boronic acid

  • MDL number MFCD11226596
  • Molecular Weight : 190.007

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C18924
Availability details for
Quantity
  • Availability
  • Add to Cart