MAN238928693 Manchester Organics

3-Methoxyphenylglyoxal hydrate

  • MDL number MFCD06800953
  • Cas Number : 32025-65-3
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-methoxyphenyl)(oxo)acetaldehyde hydrate
Inchi Code 1S/C9H8O3.H2O/c1-12-8-4-2-3-7(5-8)9(11)6-10;/h2-6H,1H3;1H2
Inchi Key LDMLATLMTDGBHP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/G10408
Availability details for
Quantity
  • Availability
  • Add to Cart