MAN238928696 Manchester Organics

4-Amino-2-chloro-3-fluorobenzonitrile

  • MDL number MFCD06808834
  • Cas Number : 757247-99-7
  • Molecular Weight : 170.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/D18925
Availability details for
Quantity
  • Availability
  • Add to Cart