MAN238928712 Manchester Organics

2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine hydrochloride

  • MDL number MFCD09999262
  • Molecular Weight : 227.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A10285

Safety Information

MSDS https://www.manchesterorganics.com/A10285
Availability details for
Quantity
  • Availability
  • Add to Cart