MAN238928714 Manchester Organics

(1H-Indazol-5-yl)methanol

  • MDL number MFCD04038636
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A15207

Safety Information

MSDS https://www.manchesterorganics.com/A15207
Availability details for
Quantity
  • Availability
  • Add to Cart