MAN238928720 Manchester Organics

1-Methyl-6-nitro-1H-indazole

  • MDL number MFCD00456124
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B18877

Safety Information

MSDS https://www.manchesterorganics.com/B18877
Availability details for
Quantity
  • Availability
  • Add to Cart