MAN238928722 Manchester Organics

Indazole-4-boronic acid pinacol ester

  • MDL number MFCD08059267
  • Cas Number : 885618-33-7
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
Inchi Code 1S/C13H17BN2O2/c1-12(2)13(3,4)18-14(17-12)10-6-5-7-11-9(10)8-15-16-11/h5-8H,1-4H3,(H,15,16)
Inchi Key YRPXZCWDXBNPBD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/D18854
Availability details for
Quantity
  • Availability
  • Add to Cart