MAN238928736 Manchester Organics

Ethyl 1,4-dimethyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD11226402
  • Molecular Weight : 168.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A18947
Availability details for
Quantity
  • Availability
  • Add to Cart