MAN238928747 Manchester Organics

2-Hydroxy-6-(trifluoromethyl)nicotinitrile

  • MDL number MFCD09832863
  • Cas Number : 802565-31-7
  • Molecular Weight : 188.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/E18951
Availability details for
Quantity
  • Availability
  • Add to Cart