MAN238928801 Manchester Organics

2-Cyclopentylaminoethanol

  • MDL number MFCD03423780
  • Molecular Weight : 129.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A19016

Safety Information

MSDS https://www.manchesterorganics.com/A19016
Availability details for
Quantity
  • Availability
  • Add to Cart